missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-4391348
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
175205-39-7
C8H6FNS
167.201
QVCYTKXGZLOVFA-UHFFFAOYSA-N
5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
519443
4-fluoro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)F)N=C=S
This item is not returnable. View return policy
175205-39-7
C8H6FNS
167.201
QVCYTKXGZLOVFA-UHFFFAOYSA-N
5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
519443
4-fluoro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)F)N=C=S
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methylphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'