missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylaniline, 99%, Thermo Scientific Chemicals

Catalog Number 11905211
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
367-29-3
C7H8FN
125.15
MFCD00007764
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
67774
5-fluoro-2-methylaniline
CC1=CC=C(F)C=C1N
This item is not returnable. View return policy
367-29-3
C7H8FN
125.15
MFCD00007764
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
67774
5-fluoro-2-methylaniline
CC1=CC=C(F)C=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 40.0°C
Color Purple to Brown
Boiling Point 98.0°C to 100.0°C (15.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)NH2
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 125.15
Percent Purity 99%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material 5-Fluoro-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Fluoro-2-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'