missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxypyridine-3-carboxaldehyde, 98%, Thermo Scientific™

Catalog Number 15499076
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
351410-62-3
C7H6FNO2
155.128
MFCD04972398
YAGPZRLCMRMSFP-UHFFFAOYSA-N
22678323
5-fluoro-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)F
This item is not returnable. View return policy
351410-62-3
C7H6FNO2
155.128
MFCD04972398
YAGPZRLCMRMSFP-UHFFFAOYSA-N
22678323
5-fluoro-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 14158282
Sensitivity Air and moisture sensitive
Synonym 5-fluoro-2-methoxynicotinaldehyde, 5-fluoro-3-formyl-2-methoxypyridine, 3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy, 5-fluoro-2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-5-fluoropyridine-3-carbaldehyde, 5-fluoro-2-methoxy-pyridine-3-carbaldehyde, 5-fluoro-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde,5-fluoro-2-methoxy, 5-fluoro-2-methoxy-3-pyridine carboxaldehyde, pubchem16100
Formula Weight 155.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Fluoro-2-methoxypyridine-3-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'