missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxyphenylboronic acid, 98%, Thermo Scientific™

Catalog Number p-4228679
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
179897-94-0
C7H8BFO3
169.95
MFCD01863526
CCQKIRUMTHHPSX-UHFFFAOYSA-N
5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
2782673
(5-fluoro-2-methoxyphenyl)boronic acid
COC1=CC=C(F)C=C1B(O)O
This item is not returnable. View return policy
179897-94-0
C7H8BFO3
169.95
MFCD01863526
CCQKIRUMTHHPSX-UHFFFAOYSA-N
5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
2782673
(5-fluoro-2-methoxyphenyl)boronic acid
COC1=CC=C(F)C=C1B(O)O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 156.0°C to 158.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 169.95
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Fluoro-2-methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'