missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62432.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487426

  • £343.40 / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
280.1
5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester

Specifications

5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
1400976-17-1
96%
5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester
1g
Solid
280.1
Videos
SDS