missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7031346
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
394-04-7
C8H7FO3
170.139
MFCD00671765
WPXFJBPJUGMYOD-UHFFFAOYSA-N
5-fluoro-2-methoxy-benzoic acid, 2-methoxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-methoxy, 5-fluoro-2-methoxylbenzoic acid, pubchem2677, 5-fluoro-o-anisic acid, 2-carboxy-4-fluoroanisole, rarechem al bo 1468, buttpark 32\01-79
2737364
5-fluoro-2-methoxybenzoic acid
COC1=C(C=C(C=C1)F)C(=O)O
This item is not returnable. View return policy
394-04-7
C8H7FO3
170.139
MFCD00671765
WPXFJBPJUGMYOD-UHFFFAOYSA-N
5-fluoro-2-methoxy-benzoic acid, 2-methoxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-methoxy, 5-fluoro-2-methoxylbenzoic acid, pubchem2677, 5-fluoro-o-anisic acid, 2-carboxy-4-fluoroanisole, rarechem al bo 1468, buttpark 32\01-79
2737364
5-fluoro-2-methoxybenzoic acid
COC1=C(C=C(C=C1)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Fluoro-3-hydroxy-2-methylbenzoic acid 1 was synthesized from 5-fluoro-2-methylbenzoic acid by nitration, reduction, diazotization and hydrolysis. Another methylcarbamoyl-substituted benzylamine 12 was synthesized from 5-fluoro-2-methylbenzoic acid via the sequential bromination,azido replacement and the reduction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 86°C to 90°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 170.14
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'