missing translation for 'onlineSavingsMsg'
Learn More

5'-Fluoro-2'-iodoacetophenone, 97%

Product Code. 15422769
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15422769

Brand: Thermo Scientific Alfa Aesar H66475.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 914225-70-0 | C8H6FIO | 264.04 g/mol

5'-Fluoro-2'-iodoacetophenone is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Fluoro-2′-iodoacetophenone is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914225-70-0
Molecular Formula C8H6FIO
Molecular Weight (g/mol) 264.04
MDL Number MFCD09033159
InChI Key NRLSRIONJVBZDT-UHFFFAOYSA-N
Synonym 5'-fluoro-2'-iodoacetophenone, 1-5-fluoro-2-iodophenyl ethanone, 2'-iodo-5'-fluoroacetophenone, 1-5-fluoro-2-iodophenyl ethan-1-one, 1-5-fluoro-2-iodo-phenyl ethanone, 5-fluoro-2-iodoacetophenone, pubchem15742, 1-5-fluoro-2-iodo-phenyl-ethanone, 1-5-fluoranyl-2-iodanyl-phenyl ethanone
PubChem CID 29934881
IUPAC Name 1-(5-fluoro-2-iodophenyl)ethanone
SMILES CC(=O)C1=C(I)C=CC(F)=C1

Specifications

Color Brown
Quantity 1 g
Beilstein 22044599
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 264.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5'-Fluoro-2'-iodoacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.