missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-2-formylbenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62170.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447116

  • £292.00 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Fluoro-2-formylbenzeneboronic acid pinacol ester
250.07
5-Fluoro-2-formylphenylboronic acid pinacol ester

Specifications

5-Fluoro-2-formylbenzeneboronic acid pinacol ester
Solid
250.07
1g
96%
5-Fluoro-2-formylphenylboronic acid pinacol ester
Videos
SDS