missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Ethyl-2-(Fmoc-amino)-4-phenylthiazole, 97%

Brand:  Alfa Aesar™ H58434.09

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15434728

  • £754.19 / 10g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Ethyl-2-(Fmoc-amino)-4-phenylthiazole
426.53
MFCD22581243

Specifications

5-Ethyl-2-(Fmoc-amino)-4-phenylthiazole
10g
97%
MFCD22581243
Pale brown
Solid
426.53
Videos
SDS