missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole, 95%

Brand:  Alfa Aesar™ H61174.06

Code : NEW

Additional Details : CAS Number : 102625-64-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15415515

  • £15.69 / 5g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole
102625-64-9
367.37
MFCD07368273

Specifications

5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole
109° to 118°C
102625-64-9
95%
MFCD07368273
Off-white
5g
Powder
367.37
Videos
SDS