missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3586374
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
305832-67-1
C5H4BNO2S
152.96
MFCD02094029
ZEOMEPSYIIQIND-UHFFFAOYSA-N
5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
4198743
(5-cyanothiophen-2-yl)boronic acid
OB(O)C1=CC=C(S1)C#N
This item is not returnable. View return policy
305832-67-1
C5H4BNO2S
152.96
MFCD02094029
ZEOMEPSYIIQIND-UHFFFAOYSA-N
5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
4198743
(5-cyanothiophen-2-yl)boronic acid
OB(O)C1=CC=C(S1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 175°C to 180°C
Color Pink to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and thf
Formula Weight 152.97
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Cyanothiophene-2-boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Cyanothiophene-2-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'