missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-4-phenyl-2-(3-pyridyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7039603
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1858252-21-7
C15H9N3S
263.318
MFCD22683027
QBABKNRVLIDXCF-UHFFFAOYSA-N
4-phenyl-2-pyridin-3-yl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-3-pyridyl thiazole
98000539
4-phenyl-2-pyridin-3-yl-1,3-thiazole-5-carbonitrile
C1=CC=C(C=C1)C2=C(SC(=N2)C3=CN=CC=C3)C#N
This item is not returnable. View return policy
1858252-21-7
C15H9N3S
263.318
MFCD22683027
QBABKNRVLIDXCF-UHFFFAOYSA-N
4-phenyl-2-pyridin-3-yl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-3-pyridyl thiazole
98000539
4-phenyl-2-pyridin-3-yl-1,3-thiazole-5-carbonitrile
C1=CC=C(C=C1)C2=C(SC(=N2)C3=CN=CC=C3)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 263.32
Percent Purity 97%
Chemical Name or Material 5-Cyano-4-phenyl-2-(3-pyridyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'