Learn More
Alfa Aesar™ 5-Cyano-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole, 95%
Brand: Alfa Aesar™ H63166.03
Code : NEW
Additional Details : CAS Number : 246028-92-2 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
5-Cyano-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole | |
280.32 | |
UN3439 | |
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)indole-5-carbonitrile |
246028-92-2 | |
MFCD20266619 | |
8925724 |
Specifications
5-Cyano-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole | |
246028-92-2 | |
95% | |
MFCD20266619 | |
8925724 | |
White to off-white |
1g | |
Solid | |
280.32 | |
UN3439 | |
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)indole-5-carbonitrile |