missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63066.MD

Code : NEW

Additional Details : CAS Number : 127792-81-8 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15488106

  • £35.83 / 250mg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
223.27
UN3439
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole-5-carbonitrile
127792-81-8
MFCD26741993
4189205

Specifications

5-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
250mg
Solid
223.27
UN3439
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole-5-carbonitrile
Off-white to yellow
127792-81-8
97%
MFCD26741993
4189205
Videos
SDS