missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Cyano-2-fluoropyridine, 95%

Brand:  Alfa Aesar™ H64278.03

Code : NEW

Additional Details : CAS Number : 3939-12-6 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15461399

  • £41.50 / 1g


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

5-Cyano-2-fluoropyridine
122.1
UN3439
2-Fluoropyridine-5-carbonitrile
3959-05-5
MFCD09261087
471855

Specifications

5-Cyano-2-fluoropyridine
51° to 52°C
3959-05-5
95%
MFCD09261087
471855
Off-white
1g
Solid
122.1
UN3439
2-Fluoropyridine-5-carbonitrile
Videos
SDS