missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5α -Cholest-8(14)-en-3β-ol-15-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | (3S,5S,9R,10S,13R,17R)-3-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,4,5,6,7,9,11,12,16,17-dodecahydrocyclopenta[a]phenanthren-15-one |
Formula Weight | 400.64 |
Percent Purity | 97% |
Chemical Name or Material | 5α -cholest-8(14)-en-3β-ol-15-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'