missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible in water.
Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
Not miscible in water.
Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
Specifications
Density | 1.623 |
Boiling Point | 117°C to 120°C (14 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.586 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 130710 |
Sensitivity | Moisture sensitive |
Solubility Information | Not miscible in water. |
Formula Weight | 217.09 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'