missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-91723
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
635-93-8
C7H5ClO2
156.57
MFCD00003331
FUGKCSRLAQKUHG-UHFFFAOYSA-N
5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
12481
5-chloro-2-hydroxybenzaldehyde
OC1=CC=C(Cl)C=C1C=O
This item is not returnable. View return policy
635-93-8
C7H5ClO2
156.57
MFCD00003331
FUGKCSRLAQKUHG-UHFFFAOYSA-N
5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
12481
5-chloro-2-hydroxybenzaldehyde
OC1=CC=C(Cl)C=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 102°C to 103°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 156.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylaldehyde, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'