missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 12113751
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
872041-85-5
C5H5BClNO2
157.36
MFCD06798243
NJXYBTMCTZAUEE-UHFFFAOYSA-N
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397
20111877
(5-chloropyridin-3-yl)boronic acid
OB(O)C1=CC(Cl)=CN=C1
This item is not returnable. View return policy
872041-85-5
C5H5BClNO2
157.36
MFCD06798243
NJXYBTMCTZAUEE-UHFFFAOYSA-N
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397
20111877
(5-chloropyridin-3-yl)boronic acid
OB(O)C1=CC(Cl)=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 157.36
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-3-boronic acid
compliance-icons
Product Identifier
  • 5-Chloropyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'