missing translation for 'onlineSavingsMsg'
Learn More

5-chloropyridazin-3(2H)-one, 97%, Maybridge

£162.00 - £643.00

Chemical Identifiers

CAS 660425-07-0
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD12198134
InChI Key NHMLZGFOEYLZEN-UHFFFAOYSA-N
Synonym 5-chloropyridazin-3-2h-one, 5-chloropyridazin-3 2h-one, 5-chloropyridazin-3-ol, 5-chloro-2h-pyridazin-3-one, 3 2h-pyridazinone, 5-chloro, acmc-20dlsu, 5-chloropyridazin-3-one, 5-chloropyridazine-3-ol, 5-chloro-pyridazin-3-one, 5-chloropyridazin-3 2h one
PubChem CID 19905718
IUPAC Name 4-chloro-1H-pyridazin-6-one
SMILES C1=C(C=NNC1=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10450574 SDS View Documents Maybridge
MO08305DA
1g Amber Glass Bottle £162.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-07-2020
Please to purchase this item. Need a web account? Register with us today!
10021724 SDS View Documents Maybridge
MO08305DE
5g Amber Glass Bottle £643.00
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

660425-07-0
130.531
NHMLZGFOEYLZEN-UHFFFAOYSA-N
19905718
C1=C(C=NNC1=O)Cl
C4H3ClN2O
MFCD12198134
5-chloropyridazin-3-2h-one, 5-chloropyridazin-3 2h-one, 5-chloropyridazin-3-ol, 5-chloro-2h-pyridazin-3-one, 3 2h-pyridazinone, 5-chloro, acmc-20dlsu, 5-chloropyridazin-3-one, 5-chloropyridazine-3-ol, 5-chloro-pyridazin-3-one, 5-chloropyridazin-3 2h one
4-chloro-1H-pyridazin-6-one

Specifications

5-chloropyridazin-3(2H)-one
1g
97.0
C4H3ClN2O
5-chloropyridazin-3-2h-one, 5-chloropyridazin-3 2h-one, 5-chloropyridazin-3-ol, 5-chloro-2h-pyridazin-3-one, 3 2h-pyridazinone, 5-chloro, acmc-20dlsu, 5-chloropyridazin-3-one, 5-chloropyridazine-3-ol, 5-chloro-pyridazin-3-one, 5-chloropyridazin-3 2h one
C1=C(C=NNC1=O)Cl
130.531
130.53
Amber Glass Bottle
660425-07-0
97%
MFCD12198134
NHMLZGFOEYLZEN-UHFFFAOYSA-N
4-chloro-1H-pyridazin-6-one
19905718
97%