missing translation for 'onlineSavingsMsg'
Learn More

5-chloropyridazin-3(2H)-one, 97%, Thermo Scientific™

Catalog Number 10450574
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
660425-07-0
C4H3ClN2O
130.53
MFCD12198134
NHMLZGFOEYLZEN-UHFFFAOYSA-N
5-chloropyridazin-3-2h-one, 5-chloropyridazin-3 2h-one, 5-chloropyridazin-3-ol, 5-chloro-2h-pyridazin-3-one, 3 2h-pyridazinone, 5-chloro, acmc-20dlsu, 5-chloropyridazin-3-one, 5-chloropyridazine-3-ol, 5-chloro-pyridazin-3-one, 5-chloropyridazin-3 2h one
19905718
4-chloro-1H-pyridazin-6-one
ClC1=CC(=O)NN=C1
This item is not returnable. View return policy
660425-07-0
C4H3ClN2O
130.53
MFCD12198134
NHMLZGFOEYLZEN-UHFFFAOYSA-N
5-chloropyridazin-3-2h-one, 5-chloropyridazin-3 2h-one, 5-chloropyridazin-3-ol, 5-chloro-2h-pyridazin-3-one, 3 2h-pyridazinone, 5-chloro, acmc-20dlsu, 5-chloropyridazin-3-one, 5-chloropyridazine-3-ol, 5-chloro-pyridazin-3-one, 5-chloropyridazin-3 2h one
19905718
4-chloro-1H-pyridazin-6-one
ClC1=CC(=O)NN=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 130.53
Percent Purity 97%
Chemical Name or Material 5-chloropyridazin-3(2H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'