missing translation for 'onlineSavingsMsg'
Learn More

5-Chloromethyl-1H-tetrazole, 95%, Thermo Scientific Chemicals

Catalog Number 15408208
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
55408-11-2
C2H3ClN4
118.52
MFCD00512845
AGWSISOYPHROLN-UHFFFAOYSA-N
5-chloromethyl-1h-tetrazole, 5-chloromethyl-1h-1,2,3,4-tetrazole, 5-chloromethyl-2h-tetrazole, 1h-tetrazole, 5-chloromethyl, 5-chloromethyl-1 2 h-tetrazole, 5-chloromethyl-2h-1,2,3,4-tetrazole, chloromethyltetrazole, pubchem9133, 5-chloromethyltetrazole
12205080
5-(chloromethyl)-2H-tetrazole
ClCC1=NNN=N1
This item is not returnable. View return policy
55408-11-2
C2H3ClN4
118.52
MFCD00512845
AGWSISOYPHROLN-UHFFFAOYSA-N
5-chloromethyl-1h-tetrazole, 5-chloromethyl-1h-1,2,3,4-tetrazole, 5-chloromethyl-2h-tetrazole, 1h-tetrazole, 5-chloromethyl, 5-chloromethyl-1 2 h-tetrazole, 5-chloromethyl-2h-1,2,3,4-tetrazole, chloromethyltetrazole, pubchem9133, 5-chloromethyltetrazole
12205080
5-(chloromethyl)-2H-tetrazole
ClCC1=NNN=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 118.53
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Chloromethyl-1H-tetrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'