missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-o-anisic acid, 99%, Thermo Scientific™

Catalog Number p-3593351
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3438-16-2
MFCD00017546
HULDRQRKKXRXBI-UHFFFAOYSA-N
5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid
76970
5-chloro-2-methoxybenzoic acid
COC1=C(C=C(C=C1)Cl)C(=O)O
This item is not returnable. View return policy
3438-16-2
MFCD00017546
HULDRQRKKXRXBI-UHFFFAOYSA-N
5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid
76970
5-chloro-2-methoxybenzoic acid
COC1=C(C=C(C=C1)Cl)C(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 98.0°C to 100.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H7ClO3
Linear Formula CH3OC6H3(Cl)CO2H
Quantity 5 g
Molecular Weight (g/mol) 186.59
Formula Weight 186.59
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-o-anisic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-2-anisic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'