missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-8-hydroxy-7-iodoquinoline, 98%

Brand:  Alfa Aesar™ A11137.09

Code : NEW

Additional Details : CAS Number : 130-26-7 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11419043

  • £18.40 / 10g


Estimated Shipment date
from Supplier 01-05-2018
Add to basket

Description

Description

Applications
Clioquinol is a metal chelating ligand which displays protective effects in various neurodegenerative states as well as being a component in antifungal and antiprotozoal agents. Clioquinol rescues lead-induced impairments of synaptic plasticity in an animal models

Solubility
Soluble in DMSO (>25 mg/ml), boiling alcohol ((1:43)), methanol, and chloroform ((1:120)).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

5-Chloro-8-hydroxy-7-iodoquinoline
C9H5ClINO
MFCD00006787
14,5031
130-26-7
305.49
153637
Clioqinol

Specifications

5-Chloro-8-hydroxy-7-iodoquinoline
10g
98%
305.49
153637
Clioqinol
≈175°C (dec.)
130-26-7
C9H5ClINO
MFCD00006787
14,5031
Videos
SDS