missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-nitroisoindoline-1,3-dione, ≥95%, Thermo Scientific™

Catalog Number p-2702952
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
6015-57-2
C8H3ClN2O4
226.572
MFCD00052331
ADLVDYMTBOSDFE-UHFFFAOYSA-N
4-chloro-5-nitrophthalimide, 5-chloro-6-nitroisoindoline-1,3-dione, 5-chloro-6-nitro-2h-isoindole-1,3-dione, acmc-1b2sd, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro
2801129
5-chloro-6-nitroisoindole-1,3-dione
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O
This item is not returnable. View return policy
6015-57-2
C8H3ClN2O4
226.572
MFCD00052331
ADLVDYMTBOSDFE-UHFFFAOYSA-N
4-chloro-5-nitrophthalimide, 5-chloro-6-nitroisoindoline-1,3-dione, 5-chloro-6-nitro-2h-isoindole-1,3-dione, acmc-1b2sd, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro
2801129
5-chloro-6-nitroisoindole-1,3-dione
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.57
Percent Purity 95%
Chemical Name or Material 5-Chloro-6-nitroisoindoline-1, 3-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'