missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-nitrothiophene-2-sulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7081695
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
58457-24-2
C4HCl2NO4S2
262.075
MFCD00067978
SGHAHFHTASPNKN-UHFFFAOYSA-N
2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
2735783
5-chloro-4-nitrothiophene-2-sulfonyl chloride
C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl
This item is not returnable. View return policy
58457-24-2
C4HCl2NO4S2
262.075
MFCD00067978
SGHAHFHTASPNKN-UHFFFAOYSA-N
2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
2735783
5-chloro-4-nitrothiophene-2-sulfonyl chloride
C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 54°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 262.09
Percent Purity 98%
Chemical Name or Material 5-Chloro-4-nitrothiophene-2-sulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'