missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-3-methylbenzo[b]thiophene, 97%, Thermo Scientific™

Catalog Number p-2702725
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
19404-18-3
C9H7ClS
182.665
MFCD00052506
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
This item is not returnable. View return policy
19404-18-3
C9H7ClS
182.665
MFCD00052506
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 182.67
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'