missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde, 97%

Brand:  Alfa Aesar™ H33065.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478295

  • £96.40 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
202.61
MFCD22682805

Specifications

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
1g
Solid
202.61
Air Sensitive
1823049-90-6
97%
MFCD22682805
Videos
SDS