missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific™

Catalog Number 15488295
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1823049-90-6
C9H8ClFO2
202.609
MFCD22682805
VILWOSMVUDTFHT-UHFFFAOYSA-N
98001124
5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
CC1=C(C(=C(C=C1C=O)Cl)OC)F
This item is not returnable. View return policy
1823049-90-6
C9H8ClFO2
202.609
MFCD22682805
VILWOSMVUDTFHT-UHFFFAOYSA-N
98001124
5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
CC1=C(C(=C(C=C1C=O)Cl)OC)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 202.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'