missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-nitrobenzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 329760050

Code : 29

Additional Details : CAS Number : 6628-86-0 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11379906

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

5-Chloro-2-nitrobenzaldehyde
96.0
97%
C7H4ClNO3
185.57
6628-86-0
100.0
97%
ClC6H3(NO2)CHO
MFCD00007289

Specifications

5-Chloro-2-nitrobenzaldehyde
96.0
100.0
77.7° to 78.4°C
Yellow
97%
C7H4ClNO3
185.57
6628-86-0
Authentic
Glass bottle
5g
Powder
97%
ClC6H3(NO2)CHO
MFCD00007289
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Chloro-2-nitrobenzaldehyde

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS