missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-nitro-4-(trifluoromethyl)aniline, 95%

Brand:  Alfa Aesar™ H64456.03

Code : NEW

Additional Details : CAS Number : 35375-74-7 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15451669

  • £91.69 / 1g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-nitro-4-(trifluoromethyl)aniline
240.57
UN2811
35375-74-7
MFCD01312277

Specifications

5-Chloro-2-nitro-4-(trifluoromethyl)aniline
112° to 114°C
35375-74-7
95%
MFCD01312277
Yellow
1g
Solid
240.57
UN2811
Videos
SDS