missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-chloro-2-(methylthio)aniline, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | NH2C6H3(Cl)SCH3 |
Quantity | 1 g |
Synonym | 5-chloro-2-methylthio aniline, 5-chloro-2-methylsulfanyl aniline, 5-chloro-2-methylthiophenylamine, 5-chloro-2-methylthio benzenamine, 5-chloro-2-methylsulfanyl-phenylamine, benzenamine,5-chloro-2-methylthio, benzenamine, 5-chloro-2-methylthio |
Formula Weight | 173.67 |
Percent Purity | 95% |
Chemical Name or Material | 5-Chloro-2-(methylthio)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'