Learn More
Alfa Aesar™ 5-Chloro-2-methoxypyridine, 98%
Brand: Alfa Aesar™ H59376.04
Code : NEW
Additional Details : CAS Number : 13473-01-3 Weight : 0.00200kg Transport : UN number : 1993 Chem class : 1 Pack group : III
Quantity | 2g |
---|
Chemical Identifiers
5-Chloro-2-methoxypyridine | |
143.57 | |
UN1993 |
13473-01-3 | |
MFCD06254388 |
Specifications
5-Chloro-2-methoxypyridine | |
181° to 182°C | |
2g | |
1.5260 | |
98% | |
MFCD06254388 |
1.193g/mL | |
Colorless to pale yellow | |
13473-01-3 | |
Liquid | |
143.57 | |
UN1993 |