missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-methoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. p-4228518 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10724112

Brand: Thermo Scientific Acros 434480010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 89694-48-4
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD01318966
InChI Key FMBVAOHFMSQDGT-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid
PubChem CID 2735751
IUPAC Name (5-chloro-2-methoxyphenyl)boronic acid
SMILES COC1=C(C=C(Cl)C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 141°C to 143°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186.4
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-chloro-2-methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-2-methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.