missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-methoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65899.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429986

  • £74.73 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-methoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
315.8
MFCD04215662

Specifications

5-Chloro-2-methoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
Solid
315.8
1g
95%
MFCD04215662
Videos
SDS