missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-iodopyridine, 98%, Thermo Scientific™

Catalog Number 15491499
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
244221-57-6
C5H3ClIN
239.44
MFCD07368864
CXWLXKZIXLOBCC-UHFFFAOYSA-N
2-iodo-5-chloropyridine, 5-chloro-2-iodo-pyridine, pubchem6574, acmc-209gc5, ksc493k7f, pyridine, 5-chloro-2-iodo
11118148
5-chloro-2-iodopyridine
C1=CC(=NC=C1Cl)I
This item is not returnable. View return policy
244221-57-6
C5H3ClIN
239.44
MFCD07368864
CXWLXKZIXLOBCC-UHFFFAOYSA-N
2-iodo-5-chloropyridine, 5-chloro-2-iodo-pyridine, pubchem6574, acmc-209gc5, ksc493k7f, pyridine, 5-chloro-2-iodo
11118148
5-chloro-2-iodopyridine
C1=CC(=NC=C1Cl)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is applied in an efficient preparation of indolizines through a tandem palladium-catalyzed cross-coupling reaction and cycloisomerization and also in the synthesis, and structure-activity relationships of 2-aminoquinazoline derivatives as a novel class of metabotropic glutamate receptor 5 negative allosteric modulators.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 9321405
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 239.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Chloro-2-iodopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'