missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxy-3-methylbenzaldehyde, 95%, Thermo Scientific™

Catalog Number p-2701453
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
23602-63-3
C8H7ClO2
170.592
MFCD00275687
NSKZAOKQZDLHGO-UHFFFAOYSA-N
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
2774358
5-chloro-2-hydroxy-3-methylbenzaldehyde
CC1=C(C(=CC(=C1)Cl)C=O)O
This item is not returnable. View return policy
23602-63-3
C8H7ClO2
170.592
MFCD00275687
NSKZAOKQZDLHGO-UHFFFAOYSA-N
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
2774358
5-chloro-2-hydroxy-3-methylbenzaldehyde
CC1=C(C(=CC(=C1)Cl)C=O)O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 170.59
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'