missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(5-methyl-2-furyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64584.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401869

  • £230.02 / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(5-methyl-2-furyl)benzimidazole
232.67
MFCD20390386

Specifications

5-Chloro-2-(5-methyl-2-furyl)benzimidazole
95%
232.67
Solid
5g
MFCD20390386
Videos
SDS