missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(4-nitrophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64228.03

Code : NEW

Additional Details : CAS Number : 46992-58-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471329

  • £119.78 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(4-nitrophenyl)benzimidazole
46992-58-9
273.68
MFCD15173047

Specifications

5-Chloro-2-(4-nitrophenyl)benzimidazole
46992-58-9
95%
MFCD15173047
1g
Solid
273.68
Videos
SDS