missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(4-isopropylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64491.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431729

  • £351.92 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(4-isopropylphenyl)benzimidazole
270.76
MFCD05042202

Specifications

5-Chloro-2-(4-isopropylphenyl)benzimidazole
Solid
270.76
5g
95%
MFCD05042202
Videos
SDS