Alfa Aesar™ 5-Chloro-2-(4-fluorophenyl)benzimidazole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64620.06

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 21627-71-4 Weight: 0.00500kg


Quantity 5g

Product Code. 15471899

Quantity Price
1 £ 460.04 / 5g
Estimated Shipment date
from Supplier 14-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-(4-fluorophenyl)benzimidazole
CAS 21627-71-4
Formula Weight 246.67
MDL Number MFCD00220105

Specification

Formula Weight 246.67
Physical Form Solid
Chemical Name or Material 5-Chloro-2-(4-fluorophenyl)benzimidazole
Assay 95%
CAS 21627-71-4
Quantity 5g
MDL Number MFCD00220105