missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-(3-nitrophenyl)benzimidazole, 95%
Brand: Alfa Aesar™ H64951.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
5-Chloro-2-(3-nitrophenyl)benzimidazole | |
273.68 | |
MFCD15173045 |
Specifications
5-Chloro-2-(3-nitrophenyl)benzimidazole | |
Solid | |
273.68 |
1g | |
95% | |
MFCD15173045 |