missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(3-methylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64142.03

UNSPSC : 12352005

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15411219

  • £119.78 / 1g


Estimated Shipment date
from Supplier 07-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

5-Chloro-2-(3-methylphenyl)benzimidazole
242.71
MFCD18914844
5-Chloro-2-(m-tolyl)benzimidazole

Specifications

242.71
Solid
5-Chloro-2-(3-methylphenyl)benzimidazole
5-Chloro-2-(m-tolyl)benzimidazole
95%
MFCD18914844
1g
Documentation

Technical Documents