missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(3-methylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64142.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15421219

  • £597.84 / 5g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(3-methylphenyl)benzimidazole
242.71
MFCD18914844
5-Chloro-2-(m-tolyl)benzimidazole

Specifications

5-Chloro-2-(3-methylphenyl)benzimidazole
95%
242.71
5-Chloro-2-(m-tolyl)benzimidazole
Solid
5g
MFCD18914844
Videos
SDS