missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(3,4-difluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64923.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412349

  • £70.28 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(3,4-difluorophenyl)benzimidazole
264.66
MFCD20044588

Specifications

5-Chloro-2-(3,4-difluorophenyl)benzimidazole
Solid
264.66
1g
95%
MFCD20044588
Videos
SDS