missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(3,4-difluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64923.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422349

  • £351.92 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(3,4-difluorophenyl)benzimidazole
264.66
MFCD20044588

Specifications

5-Chloro-2-(3,4-difluorophenyl)benzimidazole
Solid
264.66
5g
95%
MFCD20044588
Videos
SDS