missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2-thienyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64757.06

Code : NEW

Additional Details : CAS Number : 27503-77-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15442089

  • £460.04 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(2-thienyl)benzimidazole
27503-77-1
234.7
MFCD01004488

Specifications

5-Chloro-2-(2-thienyl)benzimidazole
27503-77-1
95%
MFCD01004488
5g
Solid
234.7
Videos
SDS