missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2-methoxyphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64870.03

Code : NEW

Additional Details : CAS Number : 133688-90-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412269

  • £59.78 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(2-methoxyphenyl)benzimidazole
133688-90-1
258.7
MFCD11849214

Specifications

5-Chloro-2-(2-methoxyphenyl)benzimidazole
133688-90-1
95%
MFCD11849214
1g
Solid
258.7
Videos
SDS