missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2-methoxyphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64870.06

Code : NEW

Additional Details : CAS Number : 133688-90-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422269

  • £298.92 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(2-methoxyphenyl)benzimidazole
133688-90-1
258.7
MFCD11849214

Specifications

5-Chloro-2-(2-methoxyphenyl)benzimidazole
133688-90-1
95%
MFCD11849214
5g
Solid
258.7
Videos
SDS