missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2-chlorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64165.03

Code : NEW

Additional Details : CAS Number : 14225-75-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15481239

  • £99.64 / 1g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(2-chlorophenyl)benzimidazole
14225-75-3
263.12
MFCD01929354

Specifications

5-Chloro-2-(2-chlorophenyl)benzimidazole
14225-75-3
95%
MFCD01929354
1g
Solid
263.12
Videos
SDS